missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoisovaleryl)urea 98.0+%, TCI America™

Catalog No. B284225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B284225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B284225G Supplier TCI America Supplier No. B284225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 496-67-3
Molecular Formula C6H11BrN2O2
Molecular Weight (g/mol) 223.07
InChI Key CMCCHHWTTBEZNM-UHFFFAOYSA-N
Synonym bromisoval, bromisovalum, bromovalerylurea, bromvalerylurea, bromural, bromcarbamide, bromizoval, bromocarbamide, bromaral, bromisovalerylurea
PubChem CID 2447
ChEBI CHEBI:77043
IUPAC Name 2-bromo-N-carbamoyl-3-methylbutanamide
SMILES CC(C)C(C(=O)NC(=O)N)Br

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 25 g
Formula Weight 223.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Bromoisovaleryl)urea
EINECSNumber (9)-1139
RTECSNumber YS3150000
TSCA Yes