missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bismaleimidoethane, Thermo Scientific Chemicals

Catalog No. AC466065000
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466065000 500 mg
AC466061000 100 mg
AC466060010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC466065000 Supplier Thermo Scientific Chemicals Supplier No. 466065000
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5132-30-9
Molecular Formula C10H8N2O4
Molecular Weight (g/mol) 220.18
InChI Key PUKLCKVOVCZYKF-UHFFFAOYSA-N
IUPAC Name 1-[2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl]-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES O=C1C=CC(=O)N1CCN1C(=O)C=CC1=O

Specifications

Melting Point 195.0°C
Quantity 500 mg
Formula Weight 220.18
Chemical Name or Material 1,2-Bismaleimidoethane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only