missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trimethoxysilyl)ethane, 96%

Catalog No. AAH60850AA
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60850AA 5 mL
AAH60850AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60850AA Supplier Thermo Scientific Chemicals Supplier No. H60850AA

CAS: 18406-41-2 | C8H22O6Si2 | 270.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18406-41-2
Molecular Formula C8H22O6Si2
Molecular Weight (g/mol) 270.43
MDL Number MFCD00053672
InChI Key JCGDCINCKDQXDX-UHFFFAOYSA-N
Synonym 1,2-bis trimethoxysilyl ethane, 3,3,6,6-tetramethoxy-2,7-dioxa-3,6-disilaoctane, hexamethoxydisilylethane, 1,2-ethylenebis trimethoxysilane, dow corning x1-6145a, 2,7-dioxa-3,6-disilaoctane, 3,3,6,6-tetramethoxy, trimethoxy 2-trimethoxysilylethyl silane, 1,4-disilabutane, 1,1,1,4,4,4-hexamethoxy, acmc-209ele, ethylenebis trimethoxysilane
PubChem CID 87627
SMILES CO[Si](CC[Si](OC)(OC)OC)(OC)OC

Specifications

Color Colorless
Density 1.073 g/mL
Boiling Point 103°C to 104°C
Flash Point 109°C (228°F)
Refractive Index 1.409
Linear Formula [-CH2Si(OCH3)3]2
Quantity 5 mL
UN Number UN3382
IUPAC Name trimethoxy(2-trimethoxysilylethyl)silane
Formula Weight 270.43
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,2-Bis(trimethoxysilyl)ethane
Show More Show Less
Hazard Category H330
Hazard Statement GHS H Statement
H330
Fatal if inhaled.
Precautionary Statement P260-P271-P284-P304+P340-P310-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, N.O.S.
EINECSNumber 242-285-6
RTECSNumber JG7873000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only