missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(triethoxysilyl)ethane, 95%
Description
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.
Solubility
Soluble in alcohol, aromatic and aliphatic hydrocarbons. Hydrolyzes with water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.
Solubility
Soluble in alcohol, aromatic and aliphatic hydrocarbons. Hydrolyzes with water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Specifications
Specifications
| Boiling Point | 119°C |
| Quantity | 5 mL |
| Flash Point | 113°C (235°F) |
| Refractive Index | 1.411 |
| Density | 0.958 |
| Linear Formula | [-CH2Si(OC2H5)3]2 |
| Solubility Information | Soluble in alcohol,aromatic and aliphatic hydrocarbons. Hydrolyzes with water. |
| Formula Weight | 354.59 |
| Percent Purity | 95% |
| Chemical Name or Material | 1,2-Bis(triethoxysilyl)ethane |
Safety and Handling
| Hazard Category | H301-H312-H372 |
| Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P260-P264b-P270-P280-P301+P310-P302+P352-P312-P314-P330-P363-P501c |
| EINECSNumber | 240-212-2 |
| RTECSNumber | JG7755000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only