missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(tosyloxy)ethane 99.0+%, TCI America™

Catalog No. E043825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E043825G 25 g
E0438100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E043825G Supplier TCI America Supplier No. E043825G

Chemical Identifiers

CAS 6315-52-2
Molecular Formula C16H18O6S2
Molecular Weight (g/mol) 370.434
MDL Number MFCD00008549
InChI Key LZIPBJBQQPZLOR-UHFFFAOYSA-N
Synonym 1,2-bis tosyloxy ethane, ethylene glycol ditosylate, ethylene ditosylate, ethane-1,2-diyl bis 4-methylbenzenesulfonate, ethylene glycol di-p-toluenesulfonate, 1,2-bis p-toluenesulfonyloxy ethane, ethylene di p-toluenesulfonate, 1,2-ethanediol, bis 4-methylbenzenesulfonate, ethylene glycol bis-p-toluenesulfonate, 2-p-tolylsulfonyloxy ethyl 4-methylbenzenesulfonate
PubChem CID 228289
IUPAC Name 2-(4-methylphenyl)sulfonyloxyethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCOS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 128°C
Color White
Quantity 25 g
Formula Weight 370.43
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(tosyloxy)ethane