missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(maleimido)ethane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 195°C |
| Quantity | 100 mg |
| Formula Weight | 220.18 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2-Bis(maleimido)ethane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |