missing translation for 'onlineSavingsMsg'
Learn More

[1,2-Bis(diphenylphosphino)ethane]palladium(II) Dichloride 98.0+%, TCI America™

Catalog No. B20165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20165G 5 g
B20161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20165G Supplier TCI America Supplier No. B20165G

Chemical Identifiers

CAS 19978-61-1
Molecular Formula C26H26Cl2P2Pd+2
Molecular Weight (g/mol) 577.762
MDL Number MFCD00015702
InChI Key LDJXFZUGZASGIW-UHFFFAOYSA-N
Synonym dichloro 1,2-bis diphenylphosphino ethane palladium ii
PubChem CID 131675886
IUPAC Name 2-diphenylphosphanylethyl(diphenyl)phosphane;palladium(2+);dihydrochloride
SMILES C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Pd+2]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 575.75
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,2-Bis(diphenylphosphino)ethane]palladium(II) Dichloride