missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1,2-Bis(diphenylphosphino)ethane]nickel(II) Dichloride 96.0+%, TCI America™
Supplier: TCI America B222525G
Specifications
| [1,2-Bis(diphenylphosphino)ethane]nickel(II) Dichloride | |
| C26H24Cl2NiP2 | |
| 25 g | |
| XXECWTBMGGXMKP-UHFFFAOYSA-L | |
| [2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel | |
| 131664338 | |
| ≥96.0% (T) |
| 14647-23-5 | |
| MFCD00013313 | |
| 1,2-bis diphenylphosphino ethane nickel ii chloride | |
| Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 528.02 | |
| 528.02 | |
| Crystalline Powder |
Chemical Identifiers
| 14647-23-5 | |
| 528.02 | |
| XXECWTBMGGXMKP-UHFFFAOYSA-L | |
| 131664338 | |
| Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C26H24Cl2NiP2 | |
| MFCD00013313 | |
| 1,2-bis diphenylphosphino ethane nickel ii chloride | |
| [2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel |
Safety and Handling
TSCA : No