missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[(dimethylamino)dimethylsilyl]ethane 93.0+%, TCI America™

Catalog No. B17735G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17735G Supplier TCI America Supplier No. B17735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91166-50-6
Molecular Formula C10H28N2Si2
Molecular Weight (g/mol) 232.52
MDL Number MFCD00010649
InChI Key MRAAXSSHMOFDJR-UHFFFAOYSA-N
Synonym 1,1′C-Ethylenebis(N,N,1,1-tetramethylsilanamine)
PubChem CID 4187062
IUPAC Name 2,3,3,6,6,7-hexamethyl-2,7-diaza-3,6-disilaoctane
SMILES CN(C)[Si](C)(C)CC[Si](C)(C)N(C)C

Specifications

Color Yellow
Boiling Point 103°C
Quantity 5 g
UN Number 2924
Formula Weight 232.52
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis[(dimethylamino)dimethylsilyl]ethane [Protecting Reagent for Aromatic Primary Amines]