missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(chlorodimethylsilyl)ethane 95.0+%, TCI America™

Catalog No. p-7119205
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16885G 5 g
B168825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16885G Supplier TCI America Supplier No. B16885G
Only null left

Chemical Identifiers

CAS 13528-93-3
Molecular Formula C6H16Cl2Si2
Molecular Weight (g/mol) 215.26
MDL Number MFCD00000505
InChI Key VGQOKOYKFDUPPJ-UHFFFAOYSA-N
Synonym 1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene
PubChem CID 83552
IUPAC Name chloro[2-(chlorodimethylsilyl)ethyl]dimethylsilane
SMILES C[Si](C)(Cl)CC[Si](C)(C)Cl

Specifications

Color White
Boiling Point 198°C
Quantity 5 g
UN Number 2987
Formula Weight 215.26
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2-Bis(chlorodimethylsilyl)ethane [Protecting Reagent for Primary Amines]