missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(chlorodimethylsilyl)ethane, 96%

Catalog No. AC313260050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313260050 5 g Glass bottle
AC313260250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313260050 Supplier Thermo Scientific Chemicals Supplier No. 313260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13528-93-3 | C6H16Cl2Si2 | 215.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13528-93-3
Molecular Formula C6H16Cl2Si2
Molecular Weight (g/mol) 215.26
MDL Number MFCD00000505
InChI Key VGQOKOYKFDUPPJ-UHFFFAOYSA-N
Synonym 1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene
PubChem CID 83552
IUPAC Name chloro-[2-[chloro(dimethyl)silyl]ethyl]-dimethylsilane
SMILES C[Si](C)(Cl)CC[Si](C)(C)Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 35.0°C to 38.0°C
Boiling Point 198.0°C to 200.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula [-CH2Si(CH3)2Cl]2
Quantity 5 g
Merck Index 15, 1251
Solubility Information Solubility in water: reacts.
Formula Weight 215.27
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material 1, 2-Bis(chlorodimethylsilyl)ethane, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 236-871-0
TSCA TSCA

RUO – Research Use Only