missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(3-indenyl)ethane 98.0+%, TCI America™

Catalog No. B22815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22815G 5 g
B22811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22815G Supplier TCI America Supplier No. B22815G

Chemical Identifiers

CAS 18657-57-3
Molecular Formula C20H18
Molecular Weight (g/mol) 258.36
MDL Number MFCD00191814
InChI Key CQAQBIQKEFJNRZ-UHFFFAOYSA-N
Synonym 1,2-bis 3-indenyl ethane, 1,2-di 3-indenyl ethane, 1,2-di 1h-inden-3-yl ethane, 1h-indene, 3,3'-1,2-ethanediyl bis, 3,3'-ethylenebis 1h-indene, 1h-indene,3,3'-1,2-ethanediyl bis, 1-2-3h-inden-1-yl ethyl-3h-indene, acmc-209eoe, 3-2-3h-inden-1-yl ethyl-1h-indene, 1?2-bis 3-indenyl ethane
PubChem CID 3488033
IUPAC Name 3-[2-(1H-inden-3-yl)ethyl]-1H-indene
SMILES C(CC1=CCC2=CC=CC=C12)C1=CCC2=CC=CC=C12

Specifications

Melting Point 123°C
Color Yellow
Quantity 5 g
Formula Weight 258.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(3-indenyl)ethane