Learn More
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%
CAS: 528854-34-4 | C22H44P2 | 370.542 g/mol
Supplier: Thermo Scientific Chemicals H6001703
Description
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Air sensitive. Keep away from strong oxidizing agents.
Specifications
| 1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane | |
| 67°C to 72°C | |
| MFCD08705244 | |
| Air Sensitive | |
| Insoluble in water. | |
| CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C | |
| 370.542 | |
| 370.53 |
| 528854-34-4 | |
| C22H44P2 | |
| 1 g | |
| 1,2-bis 2s,5s-2,5-di-i-propylphospholano ethan | |
| IRVIFEWWPYKALC-GXRSIYKFSA-N | |
| (2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane | |
| 11707202 | |
| ≥97% |
Chemical Identifiers
| 528854-34-4 | |
| 370.542 | |
| IRVIFEWWPYKALC-GXRSIYKFSA-N | |
| 11707202 | |
| CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C |
| C22H44P2 | |
| MFCD08705244 | |
| 1,2-bis 2s,5s-2,5-di-i-propylphospholano ethan | |
| (2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only