missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%

Catalog No. AC435191000
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435191000 100 mg Glass bottle
AC435192500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435191000 Supplier Thermo Scientific Chemicals Supplier No. 435191000

CAS: 213343-64-7 | C30H48BF4P2Rh | 660.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 213343-64-7
Molecular Formula C30H48BF4P2Rh
Molecular Weight (g/mol) 660.37
MDL Number MFCD01862465
InChI Key DSYBEQKPSKLNMC-RTXTXWNLNA-N
Synonym s,s-et-duphos-rh, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium i tetrafluoroborate, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium l tetrafluoroborate
PubChem CID 11006841
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@H]1CC[C@H](CC)P1C1=C(C=CC=C1)P1[C@@H](CC)CC[C@@H]1CC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Orange
Elemental Analysis 52.4 to 56.8% C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
IUPAC Name λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl}-2,5-diethylphospholane tetrafluoroboranuide
Formula Weight 660.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2-Bis((2S,5S)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less

RUO – Research Use Only