missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[2-methylbenzo[b]thiophen-3-yl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene 97.0+%, TCI America™

Catalog No. b22871g
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22871G 1 g
B2287100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22871G Supplier TCI America Supplier No. B22871G
Only null left

Chemical Identifiers

CAS 137814-07-4
Molecular Formula C23H14F6S2
Molecular Weight (g/mol) 468.48
MDL Number MFCD06656279
InChI Key CNLMHUFAXSWHFA-UHFFFAOYSA-N
PubChem CID 11408746
IUPAC Name 3-[3,3,4,4,5,5-hexafluoro-2-(2-methyl-1-benzothiophen-3-yl)cyclopent-1-en-1-yl]-2-methyl-1-benzothiophene
SMILES CC1=C(C2=CC=CC=C2S1)C1=C(C2=C(C)SC3=CC=CC=C23)C(F)(F)C(F)(F)C1(F)F

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 1 g
Formula Weight 468.48
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis[2-methylbenzo[b]thiophen-3-yl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene