missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(2-chloroethoxy)ethane 98.0+%, TCI America™
Supplier: TCI America B0471500ML
Specifications
| 1,2-Bis(2-chloroethoxy)ethane | |
| Colorless | |
| C6H12Cl2O2 | |
| 500 mL | |
| 1,2-bis 2-chloroethoxy ethane, triglycol dichloride, triethylene glycol dichloride, ethane, 1,2-bis 2-chloroethoxy, 1,8-dichloro-3,6-dioxaoctane, 1,2-bis chloroethoxy ethane, bis 2-chloroethoxy ethane, di 2-chloroethyl cellosolve, 2-2-chloroethoxy ethyl 2'-chloroethyl ether, unii-28jq82m13j | |
| ClCCOCCOCCCl | |
| 187.06 | |
| 187.06 | |
| Liquid |
| 112-26-5 | |
| 235°C | |
| MFCD00000976 | |
| 2810 | |
| AGYUOJIYYGGHKV-UHFFFAOYSA-N | |
| 1-chloro-2-[2-(2-chloroethoxy)ethoxy]ethane | |
| 8171 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 112-26-5 | |
| 187.06 | |
| AGYUOJIYYGGHKV-UHFFFAOYSA-N | |
| 8171 | |
| ClCCOCCOCCCl |
| C6H12Cl2O2 | |
| MFCD00000976 | |
| 1,2-bis 2-chloroethoxy ethane, triglycol dichloride, triethylene glycol dichloride, ethane, 1,2-bis 2-chloroethoxy, 1,8-dichloro-3,6-dioxaoctane, 1,2-bis chloroethoxy ethane, bis 2-chloroethoxy ethane, di 2-chloroethyl cellosolve, 2-2-chloroethoxy ethyl 2'-chloroethyl ether, unii-28jq82m13j | |
| 1-chloro-2-[2-(2-chloroethoxy)ethoxy]ethane |
Safety and Handling
EINECSNumber : (2)-0448
RTECSNumber : KH4900000
TSCA : Yes