missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-chloroethoxy)ethane 98.0+%, TCI America™

Catalog No. B047125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B047125ML 25 mL
B0471500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B047125ML Supplier TCI America Supplier No. B047125ML

Chemical Identifiers

CAS 112-26-5
Molecular Formula C6H12Cl2O2
Molecular Weight (g/mol) 187.06
MDL Number MFCD00000976
InChI Key AGYUOJIYYGGHKV-UHFFFAOYSA-N
Synonym 1,2-bis 2-chloroethoxy ethane, triglycol dichloride, triethylene glycol dichloride, ethane, 1,2-bis 2-chloroethoxy, 1,8-dichloro-3,6-dioxaoctane, 1,2-bis chloroethoxy ethane, bis 2-chloroethoxy ethane, di 2-chloroethyl cellosolve, 2-2-chloroethoxy ethyl 2'-chloroethyl ether, unii-28jq82m13j
PubChem CID 8171
IUPAC Name 1-chloro-2-[2-(2-chloroethoxy)ethoxy]ethane
SMILES ClCCOCCOCCCl

Specifications

Color Colorless
Boiling Point 235°C
Quantity 25 mL
UN Number 2810
Formula Weight 187.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(2-chloroethoxy)ethane
EINECSNumber (2)-0448
RTECSNumber KH4900000
TSCA Yes