missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium tetrafluoroborate, 99%

Catalog No. AC221610250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221610250 25 g Glass bottle
AC221611000 100 g Glass bottle
AC221615000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221610250 Supplier Thermo Scientific Chemicals Supplier No. 221610250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 429-06-1 | C8H20BF4N | 217.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 429-06-1
Molecular Formula C8H20BF4N
Molecular Weight (g/mol) 217.06
MDL Number MFCD00011827
InChI Key XJRAKUDXACGCHA-UHFFFAOYSA-N
Synonym tetraethylammonium tetrafluoroborate, tetraethylazanium tetrafluoroborate, tetraethylammoniumtetrafluoroborate, acmc-1ago3, ksc238k1t, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-1:1, tetraethyl ammonium tetrafluoroborate, tetraethylammonium tetra fluoro borate
PubChem CID 2724277
SMILES F[B-](F)(F)F.CC[N+](CC)(CC)CC

Specifications

Melting Point >300°C
Color White
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C2H5)4NBF4
Quantity 25 g
Solubility Information (4% in acetonitrile) Clear to almost clear colorless
IUPAC Name tetraethylazanium;tetrafluoroborate
Formula Weight 217.06
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetraethylammonium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 207-055-1

RUO – Research Use Only