missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%

Catalog No. AC401650010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401650010 1 g Glass bottle
AC401650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401650010 Supplier Thermo Scientific Chemicals Supplier No. 401650010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 336624-09-0 | C22H20N2Na4O10 | 564.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 336624-09-0
Molecular Formula C22H20N2Na4O10
Molecular Weight (g/mol) 564.37
MDL Number MFCD02683087
InChI Key ZWSMLJACYSGFKD-UHFFFAOYSA-J
Synonym BAPTA tetrasodium salt
IUPAC Name tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=C(OCCOC2=C(C=CC=C2)N(CC([O-])=O)CC([O-])=O)C=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Complexometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 564.36
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1,2-Bis(2-aminophenoxy)-ethane-N,N,N′,N′-tetraacetic acid, tetrasodium salt hydrate, 95%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement MOISTURE SENSITIVE
TSCA TSCA

RUO – Research Use Only