missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine Dihydrochloride 98.0+%, TCI America™

Catalog No. B23181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23181G 1 g
B23185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23181G Supplier TCI America Supplier No. B23181G

Chemical Identifiers

CAS 1820706-16-8
Molecular Formula C20H30Cl2N2
Molecular Weight (g/mol) 369.37
MDL Number MFCD11846116
InChI Key HSFKFAWYBTVLDG-UHFFFAOYNA-N
Synonym (+/-)-1,2-Dimesitylethylenediamine Dihydrochloride, (+/-)-1,2-Diamino-1,2-dimesitylethane Dihydrochloride
PubChem CID 22999734
IUPAC Name 1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine dihydrochloride
SMILES Cl.Cl.CC1=CC(C)=C(C(N)C(N)C2=C(C)C=C(C)C=C2C)C(C)=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 369.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine Dihydrochloride