missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisoxazol-3-amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 36216-80-5
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.14
InChI Key NLMVYUBGWZWUGB-UHFFFAOYSA-N
Synonym 1,2-benzisoxazol-3-amine, benzo d isoxazol-3-amine, 3-amino-1,2-benzisoxazole, benzo d isoxazol-3-ylamine, 1,2-benzoisoxazol-3-amine, benzo d isoxazole-3-ylamine, aminobenzisoxazole, amino-benzoisoxazole, zlchem 93, 3-aminobenzisoxazole
PubChem CID 2779749
IUPAC Name 1,2-benzoxazol-3-amine
SMILES C1=CC=C2C(=C1)C(=NO2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC451710050
SDS View Documents
Thermo Scientific Chemicals
451710050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

36216-80-5
134.14
1,2-benzisoxazol-3-amine, benzo d isoxazol-3-amine, 3-amino-1,2-benzisoxazole, benzo d isoxazol-3-ylamine, 1,2-benzoisoxazol-3-amine, benzo d isoxazole-3-ylamine, aminobenzisoxazole, amino-benzoisoxazole, zlchem 93, 3-aminobenzisoxazole
1,2-benzoxazol-3-amine
C7H6N2O
NLMVYUBGWZWUGB-UHFFFAOYSA-N
2779749
C1=CC=C2C(=C1)C(=NO2)N

Specifications

36216-80-5
Glass bottle
5g
NLMVYUBGWZWUGB-UHFFFAOYSA-N
1,2-benzoxazol-3-amine
2779749
97%
96% min. (HPLC)
C7H6N2O
1,2-benzisoxazol-3-amine, benzo d isoxazol-3-amine, 3-amino-1,2-benzisoxazole, benzo d isoxazol-3-ylamine, 1,2-benzoisoxazol-3-amine, benzo d isoxazole-3-ylamine, aminobenzisoxazole, amino-benzoisoxazole, zlchem 93, 3-aminobenzisoxazole
C1=CC=C2C(=C1)C(=NO2)N
134.14
134.14
1, 2-Benzisoxazol-3-amine
Videos
SDS
Documents

Documents

RUO – Research Use Only