missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisothiazol-3-one, 97%

CAS: 2634-33-5 | C7H5NOS | 151.183 g/mol

$162.58 - $600.21

Chemical Identifiers

CAS 2634-33-5
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00127753
InChI Key DMSMPAJRVJJAGA-UHFFFAOYSA-N
Synonym 1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
PubChem CID 17520
ChEBI CHEBI:167099
IUPAC Name 1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NS2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6049206
View Documents
Thermo Scientific Chemicals
H6049206
5 g
Each for $162.58
Only null left
 
AAH6049214
View Documents
Thermo Scientific Chemicals
H6049214
25 g
Each for $600.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is an antimicrobial agent and a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide, methanol.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

2634-33-5
151.183
DMSMPAJRVJJAGA-UHFFFAOYSA-N
17520
1,2-benzothiazol-3-one
C7H5NOS
MFCD00127753
1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
CHEBI:167099
C1=CC=C2C(=C1)C(=O)NS2

Specifications

2634-33-5
C7H5NOS
5 g
1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
DMSMPAJRVJJAGA-UHFFFAOYSA-N
1,2-benzothiazol-3-one
17520
151.19
1,2-Benzisothiazol-3-one
154°C to 158°C
MFCD00127753
UN3077
Soluble in dichloromethane,dimethyl sulfoxide,methanol.
C1=CC=C2C(=C1)C(=O)NS2
151.183
CHEBI:167099
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P501c

H302-H315-H317-H318

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 220-120-9

RTECSNumber : DE4620000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only