missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzisothiazol-3(2H)-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 158°C |
| Quantity | 100 g |
| UN Number | 3077 |
| Formula Weight | 151.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2-Benzisothiazol-3(2H)-one |
Safety and Handling
| EINECSNumber | (9)-1845 |
| RTECSNumber | DE4620000 |
| TSCA | Yes |