missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisothiazol-3(2H)-one 98.0+%, TCI America™

Catalog No. B37675G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37675G Supplier TCI America Supplier No. B37675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2634-33-5
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00127753
InChI Key DMSMPAJRVJJAGA-UHFFFAOYSA-N
Synonym 1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
PubChem CID 17520
ChEBI CHEBI:167099
IUPAC Name 1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NS2

Specifications

Melting Point 158°C
Quantity 5 g
UN Number 3077
Formula Weight 151.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Benzisothiazol-3(2H)-one [for Biochemical Research]
EINECSNumber (9)-1845
RTECSNumber DE4620000
TSCA Yes
Recommended Storage Refrigerator