missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedisulfonyl Dichloride 98.0+%, TCI America™

Catalog No. B11285G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11285G 5 g
B11281G 1 g
B112825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B11285G Supplier TCI America Supplier No. B11285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6461-76-3
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.114
MDL Number MFCD00059836
InChI Key YBGQXNZTVFEKEN-UHFFFAOYSA-N
Synonym benzene-1,2-disulfonyl dichloride, 1,2-benzenedisulfonyl dichloride, o-benzenedisulfonyl chloride, 1,2-benzenedisulfonyldichloride, 1,2-benzenedisulfonyl chloride, acmc-209nnc, benzenedisulfonyl chloride, benzenedisulfonyl dichloride, benzene-1,2-disulfochloride, 1,2-benzenedisulphonyl chloride
PubChem CID 246432
IUPAC Name benzene-1,2-disulfonyl chloride
SMILES C1=CC=C(C(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 275.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Benzenedisulfonyl Dichloride