missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedisulfonyl chloride

Catalog No. AAH5037506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5037506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5037506 Supplier Thermo Scientific Chemicals Supplier No. H5037506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6461-76-3 | C6H4Cl2O4S2 | 275.114 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6461-76-3
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.114
MDL Number MFCD00059836
InChI Key YBGQXNZTVFEKEN-UHFFFAOYSA-N
Synonym benzene-1,2-disulfonyl dichloride, 1,2-benzenedisulfonyl dichloride, o-benzenedisulfonyl chloride, 1,2-benzenedisulfonyldichloride, 1,2-benzenedisulfonyl chloride, acmc-209nnc, benzenedisulfonyl chloride, benzenedisulfonyl dichloride, benzene-1,2-disulfochloride, 1,2-benzenedisulphonyl chloride
PubChem CID 246432
IUPAC Name benzene-1,2-disulfonyl chloride
SMILES C1=CC=C(C(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl

Specifications

Melting Point 140°C to 146°C
Quantity 5 g
UN Number UN3261
Formula Weight 275.13
Chemical Name or Material 1,2-Benzenedisulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only