missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedisulfonic Imide 98.0+%, TCI America™

Catalog No. B41391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41391G 1 g
B41395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41391G Supplier TCI America Supplier No. B41391G
Only null left

Chemical Identifiers

CAS 4482-01-3
Molecular Formula C6H5NO4S2
Molecular Weight (g/mol) 219.23
MDL Number MFCD07779379
InChI Key QRWQFOHBHHIZKG-UHFFFAOYSA-N
Synonym o-Benzenedisulfonimide, 1,3,2-Benzodithiazole 1,1,3,3-Tetraoxide
PubChem CID 10500043
IUPAC Name 1$l^{6},3$l^{6},2-benzodithiazole 1,1,3,3-tetraoxide
SMILES C1=CC=C2C(=C1)S(=O)(=O)NS2(=O)=O

Specifications

Melting Point 197°C
Quantity 1 g
Formula Weight 219.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Benzenedisulfonic Imide