missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenediboronic Acid Bis(pinacol) Ester 98.0+%, TCI America™

Catalog No. p-7123932
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50691G 1 g
B50695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50691G Supplier TCI America Supplier No. B50691G
Only null left

Chemical Identifiers

CAS 269410-07-3
Molecular Formula C18H28B2O4
Molecular Weight (g/mol) 330.038
InChI Key VCTMQXIOUDZIGS-UHFFFAOYSA-N
Synonym 1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,2-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
PubChem CID 21923918
IUPAC Name 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2B3OC(C(O3)(C)C)(C)C

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 1 g
Formula Weight 330.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Benzenediboronic Acid Bis(pinacol) Ester