missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)pyrrolidine, 99%

Catalog No. p-7027689
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2464506 5 g
AAB2464514 25 g
AAB2464522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2464506 Supplier Thermo Scientific Chemicals Supplier No. B2464506
Only null left

CAS: 7154-73-6 | C6H14N2 | 114.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7154-73-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00003182
InChI Key WRXNJTBODVGDRY-UHFFFAOYSA-N
Synonym 1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
PubChem CID 1344
IUPAC Name 2-pyrrolidin-1-ylethanamine
SMILES C1CCN(C1)CCN

Specifications

Density 0.904
Boiling Point 161°C
Flash Point 47°C (116°F)
Odor Amine-like
Refractive Index 1.4687
Quantity 5 g
UN Number UN2734
Beilstein 102978
Sensitivity Air sensitive; Hygroscopic
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 1-(2-Aminoethyl)pyrrolidine
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 230-509-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only