missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)piperidine 97.0+%, TCI America™

Catalog No. p-7117197
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10265ML 5 mL
A102625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10265ML Supplier TCI America Supplier No. A10265ML
Only null left

Chemical Identifiers

CAS 27578-60-5
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00006516
InChI Key CJNRGSHEMCMUOE-UHFFFAOYSA-N
Synonym n-2-aminoethyl piperidine, 1-2-aminoethyl piperidine, 2-piperidin-1-yl ethanamine, 2-piperidin-1-yl ethan-1-amine, 1-piperidineethanamine, 2-piperidinoethylamine, 2-1-piperidinyl ethanamine, 2-1-piperidinyl ethylamine, n-aminoethylpiperidine, 2-piperidino-1-ethanamine
PubChem CID 33944
IUPAC Name 2-piperidin-1-ylethanamine
SMILES C1CCN(CC1)CCN

Specifications

Color Yellow
Boiling Point 184°C
Quantity 5 mL
UN Number 2734
Formula Weight 128.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Aminoethyl)piperidine