missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)piperazine, 99%

Catalog No. AC400765000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400765000 500 g Glass bottle
AC400761000 100 g Glass bottle
AC400760025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC400765000 Supplier Thermo Scientific Chemicals Supplier No. 400765000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 140-31-8 | C6H15N3 | 129.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-31-8
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.21
MDL Number MFCD00005971
InChI Key IMUDHTPIFIBORV-UHFFFAOYSA-N
Synonym n-aminoethylpiperazine, n-2-aminoethyl piperazine, 1-2-aminoethyl piperazine, 2-piperazin-1-yl ethanamine, 1-piperazineethanamine, aminoethylpiperazine, 1-piperazineethylamine, 1-aminoethylpiperazine, piperazineethanamine, piperazine, 1-2-aminoethyl
PubChem CID 8795
IUPAC Name 2-piperazin-1-ylethanamine
SMILES NCCN1CCNCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -19.0°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 218.0°C to 222.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4990 to 1.5010
Quantity 500 g
Specific Gravity 0.98
Solubility Information Solubility in water: soluble.
Viscosity 14 mPa.s (20°C)
Formula Weight 129.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-(2-Aminoethyl)piperazine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
EINECSNumber 205-411-0
RTECSNumber TK8050000
TSCA TSCA

RUO – Research Use Only