missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)piperazine, 98%

Catalog No. p-7080575
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1015430 250 g
AAA101540B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1015430 Supplier Thermo Scientific Chemicals Supplier No. A1015430

CAS: 140-31-8 | C6H15N3 | 129.21 g/mol

1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.

Chemical Identifiers

CAS 140-31-8
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.21
MDL Number MFCD00005971
InChI Key IMUDHTPIFIBORV-UHFFFAOYSA-N
Synonym n-aminoethylpiperazine, n-2-aminoethyl piperazine, 1-2-aminoethyl piperazine, 2-piperazin-1-yl ethanamine, 1-piperazineethanamine, aminoethylpiperazine, 1-piperazineethylamine, 1-aminoethylpiperazine, piperazineethanamine, piperazine, 1-2-aminoethyl
PubChem CID 8795
IUPAC Name 2-piperazin-1-ylethanamine
SMILES NCCN1CCNCC1

Specifications

Melting Point -19°C
Density 0.983
Boiling Point 222°C
Flash Point 93°C (199°F)
Refractive Index 1.5
Quantity 250 g
UN Number UN2815
Beilstein 104363
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 129.21
Percent Purity 98%
Chemical Name or Material 1- (2-Aminoethyl)piperazine
Show More Show Less
Hazard Category H227-H302-H311-H314-H317
Hazard Statement GHS H Statement
H314-H302-H312-H317-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: N-AMINOETHYLPIPERAZINE
EINECSNumber 205-411-0
RTECSNumber TK8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only