missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)-4-methylpiperazine Hydrochloride 98.0+%, TCI America™

Catalog No. a23981g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23981G Supplier TCI America Supplier No. A23981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 646051-17-4
Molecular Formula C7H18ClN3
Molecular Weight (g/mol) 179.692
InChI Key KGJPTLVFRDJLHM-UHFFFAOYSA-N
Synonym 2-(4-Methyl-1-piperazino)ethylamine Hydrochloride
PubChem CID 17962418
IUPAC Name 2-(4-methylpiperazin-1-yl)ethanamine;hydrochloride
SMILES CN1CCN(CC1)CCN.Cl

Specifications

Color White
Quantity 1 g
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Aminoethyl)-4-methylpiperazine Hydrochloride