missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)-4-methylpiperazine, 97+%

CAS: 934-98-5 | C7H17N3 | 143.234 g/mol

Supplier:  Thermo Scientific Chemicals H5237106

 View more versions of this product

Catalog No. AAH5237106


Only null left
Explore available promotions

Description

Description

1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications

Specifications

1-(2-Aminoethyl)-4-methylpiperazine
C7H17N3
5 g
Air Sensitive
GOWUDHPKGOIDIX-UHFFFAOYSA-N
2-(4-methylpiperazin-1-yl)ethanamine
70284
143.43
934-98-5
MFCD03701701
UN2735
2-4-methylpiperazin-1-yl ethanamine, 4-methyl-1-piperazineethanamine, 2-4-methylpiperazin-1-yl ethylamine, 1-piperazineethanamine, 4-methyl, 2-4-methyl-piperazin-1-yl-ethylamine, 4-methylpiperazine-1-ethylamine, 2-4-methylpiperazin-1-yl ethan-1-amine, 1-2-aminoethyl-4-methyl-piperazine, 1-2-aminoethyl-4-methylpiperazine, 1-n-aminoethyl-4-methyl piperazine
CN1CCN(CC1)CCN
143.234
CHEBI:84290
≥97%

Chemical Identifiers

934-98-5
143.234
GOWUDHPKGOIDIX-UHFFFAOYSA-N
70284
2-(4-methylpiperazin-1-yl)ethanamine
C7H17N3
MFCD03701701
2-4-methylpiperazin-1-yl ethanamine, 4-methyl-1-piperazineethanamine, 2-4-methylpiperazin-1-yl ethylamine, 1-piperazineethanamine, 4-methyl, 2-4-methyl-piperazin-1-yl-ethylamine, 4-methylpiperazine-1-ethylamine, 2-4-methylpiperazin-1-yl ethan-1-amine, 1-2-aminoethyl-4-methyl-piperazine, 1-2-aminoethyl-4-methylpiperazine, 1-n-aminoethyl-4-methyl piperazine
CHEBI:84290
CN1CCN(CC1)CCN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H312-H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 213-296-3

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only