missing translation for 'onlineSavingsMsg'
Learn More

1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals

Catalog No. AAL0676809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0676809 10 g
AAL0676818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0676809 Supplier Thermo Scientific Chemicals Supplier No. L0676809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2426-07-5 | Molecular Formula: C8H14O2 | Molecular Weight: 142.20 g/mol

1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water (partly miscible), acetone and heptane.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2426-07-5
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00005155
InChI Key LFKLPJRVSHJZPL-UHFFFAOYNA-N
Synonym 1,2,7,8-diepoxyoctane, 1,2:7,8-diepoxyoctane, 1,7-octadiene diepoxide, oxirane, 2,2'-1,4-butanediyl bis, 1,2-epoxy-7,8-epoxyoctane, 2,2'-1,4-butanediyl bisoxirane, 1,4-di oxiran-2-yl butane, octane, 1,2:7,8-diepoxy, 2-4-oxiran-2-yl butyl oxirane, ccris 963
PubChem CID 17048
ChEBI CHEBI:23705
IUPAC Name 2-[4-(oxiran-2-yl)butyl]oxirane
SMILES C1C(O1)CCCCC2CO2

Specifications

Density 0.997
Boiling Point 238°C to 240°C
Flash Point 97°C (206°F)
Refractive Index 1.445
Quantity 10 g
UN Number UN2922
Beilstein 104873
Solubility Information Soluble in water (partly miscible),acetone and heptane.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material 1,2,7,8-Diepoxyoctane
Show More Show Less
Hazard Category H302-H311-H340-H350
Hazard Statement GHS H Statement
H311-H314-H318-H341-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 219-375-9
RTECSNumber RG9450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only