missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,6-Hexanetriol 96.0+%, TCI America™
$66.46 - $537.31
Chemical Identifiers
| CAS | 106-69-4 |
|---|---|
| Molecular Formula | C6H14O3 |
| Molecular Weight (g/mol) | 134.18 |
| MDL Number | MFCD00002976 |
| InChI Key | ZWVMLYRJXORSEP-UHFFFAOYNA-N |
| Synonym | 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224 |
| PubChem CID | 7823 |
| IUPAC Name | hexane-1,2,6-triol |
| SMILES | OCCCCC(O)CO |
Chemical Identifiers
| C6H14O3 | |
| MFCD00002976 | |
| 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224 | |
| hexane-1,2,6-triol |
Specifications
| 106-69-4 | |
| 4.5 to 7.5 (300g/L H2O 20°C) | |
| C6H14O3 | |
| 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224 | |
| OCCCCC(O)CO | |
| 134.18 | |
| 134.18 | |
| Liquid |
| Yellow | |
| 178°C | |
| MFCD00002976 | |
| ZWVMLYRJXORSEP-UHFFFAOYNA-N | |
| hexane-1,2,6-triol | |
| 7823 | |
| ≥96.0% (GC) | |
| 1,2,6-Hexanetriol |
Safety and Handling
EINECSNumber : (2)-0244
RTECSNumber : MO4650000
TSCA : Yes