missing translation for 'onlineSavingsMsg'
Learn More

1,2,6-Hexanetriol, 97+%, extra pure

Catalog No. p-142328
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139831000 100 g Glass bottle
AC139835000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139831000 Supplier Thermo Scientific Chemicals Supplier No. 139831000
Only null left

CAS: 106-69-4 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-69-4
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002976
InChI Key ZWVMLYRJXORSEP-UHFFFAOYNA-N
Synonym 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
PubChem CID 7823
IUPAC Name hexane-1,2,6-triol
SMILES OCCCCC(O)CO

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 25.0°C to 32.0°C
Color Yellow
Density 1.1090g/mL
pH 4.0 to 7.0 (30% soln. at 20°C)
Boiling Point 178.0°C (6.7 mmHg)
Flash Point 198°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.4770
Linear Formula HO(CH2)4CH(OH)CH2OH
Quantity 100 g
Beilstein 01,IV,2784
Specific Gravity 1.109
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with acetone and alcohol, immiscible with benzene, toluene, heptane and, diethyl ether, practically insoluble in ether, low solubility in non-polar solvents
Formula Weight 134.18
Percent Purity 97+%
Grade Extra Pure
Physical Form Viscous Liquid
Chemical Name or Material 1, 2, 6-Hexanetriol, 97%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 203-424-6
RTECSNumber MO4650000
TSCA TSCA
Recommended Storage the product may solidify at ca. 32°C

RUO – Research Use Only