missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2:5,6-Di-O-isopropylidene-D-mannitol 97.0+%, TCI America™
Supplier: TCI America D202425G
Specifications
| 1,2:5,6-Di-O-isopropylidene-D-mannitol | |
| 120°C | |
| C12H22O6 | |
| 25 g | |
| ODYBCPSCYHAGHA-ZYUZMQFOSA-N | |
| (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol | |
| 96011 | |
| ≥97.0% (GC) |
| 1707-77-3 | |
| White-Yellow | |
| MFCD00003211 | |
| 1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213 | |
| CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C | |
| 262.302 | |
| 262.30 | |
| Crystalline Powder |
Chemical Identifiers
| 1707-77-3 | |
| 262.302 | |
| ODYBCPSCYHAGHA-ZYUZMQFOSA-N | |
| 96011 | |
| CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C |
| C12H22O6 | |
| MFCD00003211 | |
| 1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213 | |
| (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol |
Safety and Handling
TSCA : No