missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose 97.0+%, TCI America™

Catalog No. p-7127031
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D194910G 10 g
D194925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D194910G Supplier TCI America Supplier No. D194910G

Chemical Identifiers

CAS 582-52-5
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00005544
InChI Key KEJGAYKWRDILTF-MQIGXGKASA-N
Synonym 1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol
PubChem CID 40469256
IUPAC Name (3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

Melting Point 110°C
Color White
Quantity 10 g
Formula Weight 260.29
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose
RTECSNumber LZ4958000
TSCA Yes