missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose 98.0+%, TCI America™

Catalog No. D22651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22651G 1 g
D22655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22651G Supplier TCI America Supplier No. D22651G

Chemical Identifiers

CAS 2595-05-3
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.29
MDL Number MFCD00135634
InChI Key KEJGAYKWRDILTF-VVULQXIFSA-N
Synonym 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2:5,6-bis-o-1-methylethylidene, 5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose
PubChem CID 7157054
IUPAC Name (3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

Melting Point 75°C
Color White
Quantity 1 g
Formula Weight 260.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
TSCA No
Recommended Storage Freezer