missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichlorobenzene, ReagentPlus™, ≥99%, Honeywell™
Reagent Grade,≥99%
$278.35 - $278.35
Chemical Identifiers
| CAS | 120-82-1 |
|---|---|
| Molecular Formula | C6H3Cl3 |
| Molecular Weight (g/mol) | 181.44 |
| MDL Number | MFCD00000547 |
| InChI Key | PBKONEOXTCPAFI-UHFFFAOYSA-N |
| Synonym | benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene |
| PubChem CID | 13 |
| ChEBI | CHEBI:28222 |
| IUPAC Name | 1,2,4-trichlorobenzene |
| SMILES | C1=CC(=C(C=C1Cl)Cl)Cl |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 120-82-1 | |
| 181.44 | |
| PBKONEOXTCPAFI-UHFFFAOYSA-N | |
| 13 | |
| 1,2,4-trichlorobenzene |
| C6H3Cl3 | |
| MFCD00000547 | |
| benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
| CHEBI:28222 | |
| C1=CC(=C(C=C1Cl)Cl)Cl |
Specifications
| 17°C | |
| Colorless | |
| 213.5°C | |
| 6.6% (150°F) | |
| 1060°F | |
| n20/D 1.571(lit.) | |
| MFCD00000547 | |
| UN2321 | |
| benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
| C1=CC(=C(C=C1Cl)Cl)Cl | |
| 181.44 | |
| CHEBI:28222 | |
| ≥99% | |
| 1,2,4-Trichlorobenzene |
| 120-82-1 | |
| 1.454g/mL at 25°C | |
| 2.5 L | |
| 110°C (230°F) | |
| Glass Bottle | |
| C6H3Cl3 | |
| 1mmHg (40°C) | |
| 956819 | |
| PBKONEOXTCPAFI-UHFFFAOYSA-N | |
| 1,2,4-trichlorobenzene | |
| 13 | |
| 181.45g/mol | |
| ReagentPlus |
Safety and Handling
P301 + P312 + P330
H302-H315-H410
DOTInformation : 6.1/PGIII
EINECSNumber : 204-428-0
RTECSNumber : DC2100000