missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichlorobenzene (HPLC), Fisher Chemical
Supplier: Thermo Fisher Scientific O48464
Specifications
| 1, 2, 4-Trichlorobenzene | |
| 120-82-1 | |
| 214°C | |
| Amber Glass, EcoSafPak™ | |
| 96% min by GC | |
| MFCD00000547 | |
| 2321 | |
| PBKONEOXTCPAFI-UHFFFAOYSA-N | |
| 1,2,4-trichlorobenzene | |
| 13 | |
| 181.45 | |
| HPLC |
| 16°C | |
| 1.450g/cm³ | |
| 4 L | |
| Liquid | |
| C6H3Cl3 | |
| 2 hPa at 50°C | |
| benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
| C1=CC(=C(C=C1Cl)Cl)Cl | |
| 181.44 | |
| CHEBI:28222 | |
| ≥96% |
Chemical Identifiers
| 120-82-1 | |
| 181.44 | |
| PBKONEOXTCPAFI-UHFFFAOYSA-N | |
| 13 | |
| 1,2,4-trichlorobenzene |
| C6H3Cl3 | |
| MFCD00000547 | |
| benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
| CHEBI:28222 | |
| C1=CC(=C(C=C1Cl)Cl)Cl |
Safety and Handling