missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one 98.0+%, TCI America™

Catalog No. T166725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T166725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T166725G Supplier TCI America Supplier No. T166725G

Chemical Identifiers

CAS 6969-71-7
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.126
MDL Number MFCD00022632
InChI Key LJRXNXBFJXXRNQ-UHFFFAOYSA-N
Synonym 1,2,4-triazolo 4,3-a pyridin-3 2h-one, 1,2,4 triazolo 4,3-a pyridin-3 2h-one, 2h-1,2,4 triazolo 4,3-a pyridin-3-one, 1,2,4 triazolo 4,3-a pyridin-3-ol, s-triazolo 4,3-a pyridin-3-ol, 2h,3h-1,2,4 triazolo 4,3-a pyridin-3-one, 3-hydroxytriazolo 4,3-a pyridine, 1,2,4-triazolo-4,3,a pyridin-3 2h-one, 4-hydro-2h-1,2,4-triazolino 4,3-a pyridin-3-one
PubChem CID 81431
IUPAC Name 2H-[1,2,4]triazolo[4,3-a]pyridin-3-one
SMILES C1=CC2=NNC(=O)N2C=C1

Specifications

Melting Point 231°C
Color White-Yellow
Quantity 25 g
Formula Weight 135.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one