missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 98+%

Catalog No. p-7047324
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1870506 5 g
AAL1870514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1870506 Supplier Thermo Scientific Chemicals Supplier No. L1870506

CAS: 6969-71-7 | C6H5N3O | 135.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6969-71-7
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.126
MDL Number MFCD00022632
InChI Key LJRXNXBFJXXRNQ-UHFFFAOYSA-N
Synonym 1,2,4-triazolo 4,3-a pyridin-3 2h-one, 1,2,4 triazolo 4,3-a pyridin-3 2h-one, 2h-1,2,4 triazolo 4,3-a pyridin-3-one, 1,2,4 triazolo 4,3-a pyridin-3-ol, s-triazolo 4,3-a pyridin-3-ol, 2h,3h-1,2,4 triazolo 4,3-a pyridin-3-one, 3-hydroxytriazolo 4,3-a pyridine, 1,2,4-triazolo-4,3,a pyridin-3 2h-one, 4-hydro-2h-1,2,4-triazolino 4,3-a pyridin-3-one
PubChem CID 81431
IUPAC Name 2H-[1,2,4]triazolo[4,3-a]pyridin-3-one
SMILES C1=CC2=NNC(=O)N2C=C1

Specifications

Melting Point 233°C to 235°C
Quantity 5 g
Beilstein 607433
Formula Weight 135.13
Percent Purity ≥98%
Chemical Name or Material 1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-191-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only