missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazole 99.0+%, TCI America™

Supplier:  TCI America T0340100G

 View more versions of this product

Catalog No. T0340100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,2,4-Triazole
121°C
260°C
MFCD00005228
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van
C1=NC=NN1
69.067
CHEBI:35550
≥99.0% (GC)
288-88-0
White-Yellow
C2H3N3
100 g
NSPMIYGKQJPBQR-UHFFFAOYSA-N
1H-1,2,4-triazole
9257
69.07
Crystalline Powder

Chemical Identifiers

288-88-0
69.067
NSPMIYGKQJPBQR-UHFFFAOYSA-N
9257
1H-1,2,4-triazole
C2H3N3
MFCD00005228
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van
CHEBI:35550
C1=NC=NN1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5776

RTECSNumber : XZ3806000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions