missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazole 99.0+%, TCI America™

Catalog No. T0340500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0340500G 500 g
T034025G 25 g
T0340100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0340500G Supplier TCI America Supplier No. T0340500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 288-88-0
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.067
MDL Number MFCD00005228
InChI Key NSPMIYGKQJPBQR-UHFFFAOYSA-N
Synonym 1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van
PubChem CID 9257
ChEBI CHEBI:35550
IUPAC Name 1H-1,2,4-triazole
SMILES C1=NC=NN1

Specifications

Melting Point 121°C
Color White-Yellow
Boiling Point 260°C
Quantity 500 g
Formula Weight 69.07
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triazole
EINECSNumber (5)-5776
RTECSNumber XZ3806000
TSCA Yes