missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazole, 99%

Catalog No. p-7081571
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1159714 25 g
AAA1159722 100 g
AAA1159736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1159714 Supplier Thermo Scientific Chemicals Supplier No. A1159714
Only null left

CAS: 288-88-0 | C2H3N3 | 69.067 g/mol

1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and bases.

Chemical Identifiers

CAS 288-88-0
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.067
MDL Number MFCD00005228
InChI Key NSPMIYGKQJPBQR-UHFFFAOYSA-N
Synonym 1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van
PubChem CID 9257
ChEBI CHEBI:35550
IUPAC Name 1H-1,2,4-triazole
SMILES C1=NC=NN1

Specifications

Melting Point 118°C to 122°C
Boiling Point 260°C
Flash Point 158°C (316°F)
Quantity 25 g
Beilstein 104767
Merck Index 14,9605
Solubility Information Soluble in water.
Formula Weight 69.07
Percent Purity 99%
Chemical Name or Material 1,2,4-Triazole
Hazard Category H302-H319-H361d
Hazard Statement GHS H Statement
H361-H302-H319
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
EINECSNumber 206-022-9
RTECSNumber XZ3806000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only