missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic anhydride, 97%

CAS: 552-30-7 | C9H4O5 | 192.13 g/mol

$154.02 - $154.02

Chemical Identifiers

CAS 552-30-7
MDL Number MFCD00005925
InChI Key SRPWOOOHEPICQU-UHFFFAOYSA-N
Synonym trimellitic anhydride, 1,2,4-benzenetricarboxylic anhydride, anhydrotrimellic acid, trimellitic acid anhydride, 4-carboxyphthalic anhydride, tman, anhydrotrimellitic acid, trimellic acid anhydride, 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, trimellitic acid 1,2-anhydride
PubChem CID 11089
ChEBI CHEBI:53050
IUPAC Name 1,3-dioxo-2-benzofuran-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C(=O)OC2=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105370010
SDS View Documents
Thermo Scientific Chemicals
105370010
1 kg Plastic bottle
Each for $154.02
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

552-30-7
SRPWOOOHEPICQU-UHFFFAOYSA-N
11089
1,3-dioxo-2-benzofuran-5-carboxylic acid
MFCD00005925
trimellitic anhydride, 1,2,4-benzenetricarboxylic anhydride, anhydrotrimellic acid, trimellitic acid anhydride, 4-carboxyphthalic anhydride, tman, anhydrotrimellitic acid, trimellic acid anhydride, 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, trimellitic acid 1,2-anhydride
CHEBI:53050
C1=CC2=C(C=C1C(=O)O)C(=O)OC2=O

Specifications

552-30-7
100.0
White
227°C
97%
C9H4O5
1 kg
15, 9872
Solubility in water: decomposes with water. Other solubilities: 0.002g/100g ccl4- 0.06g/100g ligroin (25 c),0.4g/100g mixed xylenes- 15.5g/100g dmf (25 c),49.6g/100g acetone (25 c),21.6g/100g ethyl acetate (25 c)
C1=CC2=C(C=C1C(=O)O)C(=O)OC2=O
192.13
CHEBI:53050
97%
1, 2, 4-Benzenetricarboxylic anhydride, 97%
96.0
164.0°C to 169.0°C
390.0°C
Authentic
Plastic bottle
MFCD00005925
18, 468
trimellitic anhydride, 1,2,4-benzenetricarboxylic anhydride, anhydrotrimellic acid, trimellitic acid anhydride, 4-carboxyphthalic anhydride, tman, anhydrotrimellitic acid, trimellic acid anhydride, 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, trimellitic acid 1,2-anhydride
SRPWOOOHEPICQU-UHFFFAOYSA-N
1,3-dioxo-2-benzofuran-5-carboxylic acid
11089
192.13
Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-008-0

RTECSNumber : DC2050000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only