missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic anhydride, 97%

Catalog No. AAB2300330
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2300330 250 g
AAB23003A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2300330 Supplier Thermo Scientific Chemicals Supplier No. B2300330

CAS: 552-30-7 | C9H4O5 | 192.126 g/mol

1,2,4-Benzenetricarboxylic anhydride is used to produce good heat-proof, weather-proof and solvent-proof trimellitate plasticizers, to synthesize polyester resins, curing agent of epoxy resins, heat-proof insulated laminas, heat-proof varnishes, stabilizers, dyestuffs, pigments, surfactants. TMA can also be used as accelerant for vulcanization, treating agent of fiber and flocculant of film. It has wide application to high class aerooil, impregnating oil for power capacitors, adhesives for granules, instant adhesives, sizing agents, antismoke agents, dyestuffs, printing inks, medicines and farm chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Benzenetricarboxylic anhydride is used to produce good heat-proof, weather-proof and solvent-proof trimellitate plasticizers, to synthesize polyester resins, curing agent of epoxy resins, heat-proof insulated laminas, heat-proof varnishes, stabilizers, dyestuffs, pigments, surfactants. TMA can also be used as accelerant for vulcanization, treating agent of fiber and flocculant of film. It has wide application to high class aerooil, impregnating oil for power capacitors, adhesives for granules, instant adhesives, sizing agents, antismoke agents, dyestuffs, printing inks, medicines and farm chemicals.

Solubility
Hydrolyzes in water. Soluble in ethanol, acetone, and most other organic solvents.

Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, bases, water/ moisture.

Chemical Identifiers

CAS 552-30-7
Molecular Formula C9H4O5
Molecular Weight (g/mol) 192.126
MDL Number MFCD00005925
InChI Key SRPWOOOHEPICQU-UHFFFAOYSA-N
Synonym trimellitic anhydride, 1,2,4-benzenetricarboxylic anhydride, anhydrotrimellic acid, trimellitic acid anhydride, 4-carboxyphthalic anhydride, tman, anhydrotrimellitic acid, trimellic acid anhydride, 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, trimellitic acid 1,2-anhydride
PubChem CID 11089
ChEBI CHEBI:53050
IUPAC Name 1,3-dioxo-2-benzofuran-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C(=O)OC2=O

Specifications

Melting Point 165°C to 169°C
Density 1.54
Boiling Point 240°C to 245°C (14 mmHg)
Flash Point 227°C (440°F)
Quantity 250 g
Beilstein 9394
Sensitivity Moisture sensitive
Merck Index 14,9703
Solubility Information Hydrolyzes in water. Soluble in ethanol,acetone,and most other organic solvents.
Formula Weight 192.13
Percent Purity 97%
Chemical Name or Material 1,2,4-Benzenetricarboxylic anhydride
Show More Show Less
Hazard Category H312-H317-H318-H332-H334-H335
Hazard Statement GHS H Statement
H334-H318-H335-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P342+P311-P363-P501c
EINECSNumber 209-008-0
RTECSNumber DC2050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only