missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic acid, 98%

Catalog No. AAL0735914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0735914 25 g
AAL0735922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0735914 Supplier Thermo Scientific Chemicals Supplier No. L0735914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 528-44-9 | C9H6O6 | 210.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 528-44-9
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002470
InChI Key ARCGXLSVLAOJQL-UHFFFAOYSA-N
Synonym 1,2,4-benzenetricarboxylic acid, trimellitic acid, 1,2,4-tricarboxybenzene, 4-carboxyphthalic acid, 1,3,4-benzenetricarboxylic acid, 1,4,5-benzenetricarboxylic acid, 1,2,4-benzenetricarboxylicacid, unii-7nvy29mq5f, 7nvy29mq5f, dsstox_cid_1487
PubChem CID 10708
ChEBI CHEBI:166055
IUPAC Name benzene-1,2,4-tricarboxylic acid
SMILES OC(=O)C1=CC=C(C(O)=O)C(=C1)C(O)=O

Specifications

Melting Point ∽231°C (decomposition)
Quantity 25 g
Beilstein 2214815
Merck Index 14,9702
Formula Weight 210.14
Percent Purity 98%
Chemical Name or Material 1,2,4-Benzenetricarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-432-3
RTECSNumber DC1980000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only